NOVAX® Class 00 Electrical Safety Kits, Black Rubber Insulating Gloves
Supplier: PIP
New Product
Certificates
About this item
Electrical Safety Kits allow users to pick the class that an application requires, the color and size rubber gloves required, and Protective Industrial Products does the rest by assembling a kit that works.
- 1 pair of rubber insulating gloves (available in 11" or 14" lengths)
- 1 pair of leather protector gloves (available in 11" or 14" lengths)
- 11" or 14" nylon storage bag for gloves
Critical PPE for EV manufacturing.
Certifications: Gloves meet ASTM F696-06 (2011) standard.
Specifications
- Catalog No:
- 76944-488
- 76944-460
- 76944-486
- 76944-456
- 76944-458
- 76944-464
- 76944-462
- 76944-466
- 76944-482
- 76944-484
- 76944-490
- 76944-480
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Cuff style:
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- ASTM No.:
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a
- ASTM D120-14a