Specifications
- Description:PTS Panels® HDL cholesterol controls (Level 1 and 2)
- Storage Temperature:20...30 °C (68...86 °F)
Specifications
About this item
HDL Cholesterol Controls are intended for the quality control of PTS Diagnostic's CardioChek PA.
- Ready-to-use control materials
- Two-level control kit
- Dropper vials
PTS Panels HDL Cholesterol Controls are for professional use with the CardioChek PA and CardioChek Plus analyzers and PTS Panels test strips as quality control materials to estimate precision and to detect systematic analytical deviations of your test system. The HDL Cholesterol controls are provided in two levels.