Specifications
- Description:PTS Panels® glucose test strips
- Storage Temperature:20...30 °C (68...86 °F)
Specifications
About this item
PTS Panels Glucose test strips provide lab-quality glucose results on-site in about 45 seconds from a single fingerstick.
- Measures glucose
- 25 reflectance test strips per vial
- Room temperature storage
PTS Panels Glucose test strips are for professional use with CardioChek PA and CardioChek Plus analyzers to measure glucose for use in the diagnosis and treatment of carbohydrate metabolism disorders including diabetes mellitus, neonatal hypoglycemia, and idiopathic hypoglycemia, and of pancreatic islet cell carcinoma.
This test system will detect glucose levels from 20 to 600 mg/dl (1.11 to 33.3 mmol/L) and will display a number value for results in this range.