Specifications
- Description:PTS Panels® triglycerides test strips
- Cat. no.:77020-344
- Storage Temperature:20...30 °C (68...86 °F)
Specifications
About this item
PTS Panels Triglycerides test strips provide lab-quality triglyceride results on-site in about 45 seconds from a single fingerstick.
- Measures Triglycerides
- 25 test strips per vial – Professional
- Refrigeration not required
PTS Panels Triglycerides test strips are for professional use with CardioChek PA and CardioChek Plus analyzers to measure triglycerides in venous whole blood or capillary fingerstick blood.
Triglycerides measurements are used in the diagnosis and treatment of patients with diabetes mellitus, nephrosis, liver obstruction, other diseases involving lipid metabolism, or various endocrine disorders.
This test system will detect triglycerides levels from 50 to 500 mg/dl (0.56 to 5.65 mmol/L) and will display a number value for results in this range.