Specifications
- Description:PTS Panels® CHOL + HDL test strips
- Storage Temperature:20...30 °C (68...86 °F)
Specifications
About this item
PTS Panels CHOL+HDL test strips provide lab-quality total cholesterol and high density lipoprotein (HDL) cholesterol results on-site in as little as 45 seconds from a single fingerstick.
- Measures total cholesterol
- Calculates TC/HDL ratio
- 25 test strips per vial
- Room temperature storage
PTS Panels CHOL+HDL test strips are for professional use with CardioChek PA and CardioChek Plus analyzers to measure total cholesterol and high density lipoprotein (HDL) cholesterol in venous whole blood or capillary fingerstick blood.
Cholesterol measurements are used in the diagnosis and treatments of disorders involving excess cholesterol in the blood and lipid and lipoprotein metabolism disorders. Lipoprotein measurements are used in the diagnosis and treatment of lipid disorders (such as diabetes mellitus), atherosclerosis, and various liver and renal diseases.
This test system will display numeric results in the following ranges:
Cholesterol: 100 to 400 mg/dl (2.59 to 10.36 mmol/L)
HDL Cholesterol: 15 to 100 mg/dl (0.39 to 2.59 mmol/L)