Specifications
- Description:PTS Panels® CHOL + HDL + GLU test strips
- Storage Temperature:20...30 °C (68...86 °F)
Specifications
About this item
PTS Panels CHOL+HDL+GLU test strips provide lab-quality total cholesterol, high density lipoprotein (HDL) cholesterol, and glucose results on-site in about 90 seconds from a single fingerstick.
- Measures Total Cholesterol, HDL, and Glucose
- Calculates TC/HDL ratio
- 15 test strips per vial
- Room temperature storage
PTS Panels CHOL+HDL+GLU test strips are for professional use with CardioChek PA and CardioChek Plus analyzers to measure total cholesterol, high density lipoprotein (HDL) cholesterol, and glucose in venous whole blood or capillary fingerstick blood.
Cholesterol measurements are used in the diagnosis and treatments of disorders involving excess cholesterol in the blood and lipid and lipoprotein metabolism disorders. Lipoprotein measurements are used in the diagnosis and treatment of lipid disorders (such as diabetes mellitus), atherosclerosis, and various liver and renal diseases. Glucose measurements are used in the diagnosis and treatment of carbohydrate metabolism disorders including diabetes mellitus, neonatal hypoglycemia, and idiopathic hypoglycemia, and of pancreatic islet carcinoma.
This test system will display numeric results in the following ranges:
Cholesterol: 100 to 400 mg/dl (2.59 to 10.36 mmol/L)
HDL Cholesterol: 15 to 100 mg/dl (0.39 to 2.59 mmol/L)
Glucose: 20 to 600 mg/dl (1.11 to 33.3 mmol/L)