Order Entry
Canada
ContactUsLinkComponent
 
Bovine and Sheep Brucella Antibody IgG Titer Serologic Assay Kit
Catalog # 77007-442
Supplier:  ReVacc Scientific
CAS Number:  
undefined
Bovine and Sheep Brucella Antibody IgG Titer Serologic Assay Kit
Catalog # 77007-442
Supplier:  ReVacc Scientific
Supplier Number:  N5M98
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Bovine and sheep brucella kit
  • Cat. no.:
    77007-442

Specifications

About this item

This kit is developed for serologic test for bovine or sheep IgG titer of Anti-Brucella antibody in serum/plasma or purified bovine or sheep antibody (monoclonal and polyclonal) in vitro.

  • This kit was tested to bind to convalescent sheep sera (n = 4) with sensitivity of 100%, specificity of 75%

Assay principles: This kit is developed for a standard indirect-ELISA format, providing a rapid detection of anti-Brucella bovine or sheep IgG in serum. The kit consists of high-bind pre-coated, detachable 96-well plate, Positive control, Negative Control, an HRP-Anti-bovine or sheep IgG secondary antibody, TMB and dilution/wash/stop buffer.

Use protocol (4 simple steps):
1. Detach wells from plate based on your samples and keep at room temperature. Dilute your samples, positive and negative controls in sample dilution buffer and add them to wells (0.1 ml/well). Incubate at 37 °C for 1 hour or room temperature for 1.5 hour. Wash wells 4 times by wash solution (0.3 ml).
2. Dilute the secondary HRP antibody by wash solution and add to each well (0.1 ml). Incubate at room temperature for 0.5 to 1 hour. Avoid light by foil. Wash wells as mentioned.
3. Add TMB to each well (0.1 ml) and incubate at room temperature for 5 to 30 minutes.
4. Stop the substrate reaction by adding stop solution (0.1 ml/well). Absorbance (OD) is calculated as the absorbance at 450 nm. The OD value reflects the amount of antibody bound.

Storage and shipping condition: Shipment with room temperature. The unopened kit is stable for 6 months from the date of manufacture when stored at 2 to 8 °C. Avoid light. The shelf life is 60 days from the date of opening.