Order Entry
ContactUsLinkComponent
Swine Anti-Porcine Parvovirus Antibody IgG Titer Serologic Assay Kit (VP2)
Swine Anti-Porcine Parvovirus Antibody IgG Titer Serologic Assay Kit (VP2)
Catalog #: 77007-430
Supplier:  ReVacc Scientific
New Product
CAS Number:  
SWINE ANTI-PORCINE PARVOVIRUS KIT
Swine Anti-Porcine Parvovirus Antibody IgG Titer Serologic Assay Kit (VP2)
Catalog #: 77007-430
Supplier:  ReVacc Scientific
New Product
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Swine anti-porcine parvovirus kit
  • Cat. no.:
    77007-430

Specifications

About this item

This kit is developed for serologic test for swine IgG titer of anti-PPV structural protein VP2 in serum/plasma or purified swine antibody (monoclonal and polyclonal) in vitro.

  • This kit was tested to bind to convalescent swine sera (n = 3) with sensitivity of 80%, specificity of 92% and Cohen's kappa of 0.85 for PPV VP2 protein

Assay principles: This kit is developed for a standard indirect-ELISA format, providing a rapid detection of anti-Porcine parvovirus (PPV) IgG in serum. The kit consists of high-bind detachable 96-well plate pre-coated with indicated protein, Positive control, negative control, an HRP-Anti-Swine IgG secondary antibody, TMB and dilution/wash/stop buffer.

Use protocol (4 simple steps):
1. Detach wells from plate based on your samples and keep at room temperature. Dilute your samples, positive and negative controls in sample dilution buffer and add them to wells (0.1 ml/well). Incubate at 37 °C for 1 hour or room temperature for 1.5 hour. Wash wells 4 times by wash solution (0.3 ml).
2. Dilute the secondary antibody HRP-Anti-Swine by wash solution and add to each well (0.1 ml). Incubate at room temperature for 0.5 to 1 hour. Avoid light by foil. Wash wells as mentioned.
3. Add TMB to each well (0.1 ml) and incubate at room temperature for 5 to 30 minutes.
4. Stop the substrate reaction by adding stop solution (0.1 ml/well). Absorbance (OD) is calculated as the absorbance at 450 nm. The OD value reflects the amount of antibody bound.

Storage and shipping condition: Shipment with room temperature. The unopened kit is stable for 6 months from the date of manufacture when stored at 2 to 8 °C. Avoid light. The shelf life is 60 days from the date of opening.

Customers Who Bought This Also Bought