Specifications
- Cat. no.:76994-106
- Storage Temperature:20...30 °C (68...86 °F)
Specifications
About this item
PTS Panels Lipid allows you to get accurate lipid results in as little as 90 seconds, from one fingerstick.
- Measures Total Cholesterol, HDL, and Triglycerides
- Calculates LDL and TC/HDL ratio
- 15 test strips per vial
- Room temperature storage
PTS Panels Lipid Panel test strips are for professional use with the CardioChek PA and CardioChek Plus analyzers to measure total cholesterol, high density lipoprotein (HDL) cholesterol, and Triglycerides in venous whole blood or capillary fingerstick blood.
Cholesterol measurements are used in the diagnosis and treatments of disorders involving excess cholesterol in the blood and lipid and lipoprotein metabolism disorders. Lipoprotein measurements are used in the diagnosis and treatment of lipid disorders (such as diabetes mellitus), atherosclerosis, and various liver and renal diseases. Triglycerides measurements are used in the diagnosis and treatment of patients with diabetes mellitus, nephrosis, liver obstruction, other diseases involving lipid metabolism, or various endocrine disorders.
This test system will display numeric results in the following ranges:
Cholesterol: 100 to 400 mg/dl (2.59 to 10.36 mmol/l)
Hdl Cholesterol: 20 to 120 mg/dl (0.52 to 3.11 mmol/l)
Triglycerides: 50 to 500 mg/dl (0.57 to 5.65 mmol/l)