ContactUsLinkComponent
Replacement Air Filter for ULT Freezers
Replacement Air Filter for ULT Freezers
Catalog # 76779-128
undefined
Replacement Air Filter for ULT Freezers
Catalog # 76779-128
Supplier Number:  332297H02
Specifications
  • Description:
    Replacement air filter
  • Cat. no.:
    76779-128
  • For Use With:
    ULT chest freezers
Specifications
About this item
Replacement air filter for ultra-low temperature chest freezers.
Routine maintenance is critical for proper ultra-low temperature (ULT) freezer operation. Replacing your ULT's old air filter enables the necessary airflow to the unit's refrigeration system.