Orders LinkContactUsLinkComponent
Solution Basins, Sklar
Solution Basins, Sklar
Supplier:   Sklar
Solution Basins, Sklar
Supplier:  Sklar
Certificates
Solution Basins, Sklar
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Autoclavable and reusable solution basins made of plastic. Can be used for mixing and containing solutions, capturing fluids, or a receptacle for soiled sponges or dressings.
Product Family Options
Product Information
  • Capacity
    Material
    Dimensions
    Color
    Availability
    Price
  • 10192-806
    Solution Bassin
    Solution Bassin
    7 qt.
    Plastic
    13 5/8 x 4 5/8"
    Blue
    Discontinued
    Supplier Number: -
  • 10192-808
    Solution Bassin
    Solution Bassin
    7 qt.
    Plastic
    13 5/8 x 4 5/8"
    Blue
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $793.53
    $793.53
    New Customer Promotional Price
    Pack of 12
       $793.53
    Supplier Number: 10-1504-12
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)