To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Serum format:Frozen
- Description:FetalClone® III, US origin
- Origin:US
- Endotoxin:≤10 EU/ml
- Hemoglobin:≤25 mg/dl
- Storage temperature:<–10 °C
- Cat. no.:CA76534-650
Specifications
About this item
Origin is the most widely applicable of the FetalClone family and is designed to include fibroblasts in its applications.
- The most widely applicable of our FetalClone family of products, designed to include fibroblasts in its applications
- Offering the same basic formulation found in FetalClone II, with additional growth factors and supplements
- IgG levels comparable with those found in FBS
- Improved cell growth performance relative to FBS
- High lot-to-lot consistency
- Low endotoxin and hemoglobin levels
Conduct more cost-effective research without compromising performance. FetalClone III delivers a high culture yield at a reduced cost, plus it is readily available.
U.S. Sourced HyClone FetalClone III offers the same basic formulation of FetalClone II with additional growth factors and supplements. Sterile filtered using three sequential 100 nm (0.1 µm) pore-size rated filters. Each lot undergoes extensive quality performance and analytical testing. Additional treatments available including heat-inactivation and irradiation. Additional testing, including EMEA-conforming testing, is also available.