HyperSep™ Diol SPE Plates
Supplier: Thermo Scientific
Certificates
About this item
A short alkyl chains with polar functional groups are ideal for the isolation of polar compounds.
- Normal phase extraction of polar compounds
- Purification of polar compounds
HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
Particle size: 40 to 63 µm
Pore size: 60 Å
Surface area: 470 to 530 m²/g
Customers Who Bought This Also Bought