Order Entry
Canada
ContactUsLinkComponent
miniPCR® Agricultural Monitoring Lab: A Case Study in Antibiotic Resistance
miniPCR® Agricultural Monitoring Lab: A Case Study in Antibiotic Resistance
Catalog # 470351-990
Supplier:  AMPLYUS LLC
undefined
miniPCR® Agricultural Monitoring Lab: A Case Study in Antibiotic Resistance
Catalog # 470351-990
Supplier:  AMPLYUS LLC
Supplier Number:  KT-1010-01
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    miniPCR® agricultural monitoring lab
  • No. of Lab Groups:
    8
  • Cat. no.:
    470351-990

Specifications

About this item

Antibiotic-resistant bacteria have been traced to one farm and neighbors are concerned.

  • Techniques: Micropipetting, PCR, gel electrophoresis
  • Topics: Microbiology, infectious disease, antibiotic resistance, natural selection, environmental science, biotechnology
  • Time required: Can be completed in two 45-minute class periods or one 90-minute class period
  • Level: General high school through college

This lab (formerly known as 'miniPCR® Antibiotic Resistance Lab: Monitoring resistant organisms in the environment') presents a fictional case study of a very real problem. Epidemiologists have traced an outbreak of antibiotic-resistant bacteria to one farm. Students will use PCR to determine if the antibiotic-resistant bacteria have spread to neighboring areas. This case study introduces students to how molecular analyses can serve as a tool in environmental monitoring and surveillance. Produced in collaboration with PARE – The Prevalence of Antibiotic-Resistance in the Environment project at Tufts University. No harmful samples are used in this lab.

Reagents and consumables supplied by user:
- Agarose (electrophoresis grade)
- DNA stain (e.g., GelGreen™)
- Gel electrophoresis buffer (e.g., 1X TBE)
- Distilled or deionized H₂O (to dilute buffer concentrate – not included in Lab Companion Kit)
- Microtubes
- PCR tubes

Required equipment:
- Micropipettes and tips
- Thermal cycler
- Gel electrophoresis and visualization system

Storage notes:
- Reagents require freezer storage
- Reagents are stable for 12 months when stored in the freezer

Accessories information: The learning lab companion kit (catalog no. 470352-026) contains all the consumables (except water) needed for a class of 32 students. These reagents and consumables can also be sourced independently.
Delivery: The miniPCR® Antibiotic Resistance Lab kit contains reagents for 8 lab groups: 5X EZ PCR Master Mix, Load Ready™; PARE Primer Mix; 2 Environmental DNA samples; Negative control DNA sample; Positive control DNA sample; 100 bp DNA Ladder, Load Ready™. Note: Reagents are compatible with blueGel™ or other standard horizontal electrophoresis systems.