ContactUsLinkComponent
miniPCR® Dye Electrophoresis Lab: Microbe Hunters
miniPCR® Dye Electrophoresis Lab: Microbe Hunters
Catalog # 470352-012
Supplier:  AMPLYUS LLC
undefined
miniPCR® Dye Electrophoresis Lab: Microbe Hunters
Catalog # 470352-012
Supplier:  AMPLYUS LLC
Supplier Number:  KT-1401-01
Specifications
  • Description:
    miniPCR® dye electrophoresis lab: microbe hunters
  • No. of Lab Groups:
    8
  • Cat. no.:
    470352-012
Specifications
About this item
Astronauts aboard the International Space Station (ISS) need your help protecting themselves from disease-causing bacteria!
  • An engaging and accessible introduction to gel electrophoresis
  • Safe, non-toxic dyes that are directly visible without gel staining
  • Pre-aliquoted samples and premeasured agarose tablets for easy prep
  • Gel electrophoresis reagents included: TBE electrophoresis buffer and 8 Agarose Tabs™
  • Compatible with Bandit™, blueGel™, and traditional gel electrophoresis systems
  • A low-cost jumping-off point to explore microbiology, biotechnology, and molecular identification!
This dye electrophoresis lab offers students a close-up look at the invisible world of microbes that surrounds us. Students work as part of a team of NASA microbiologists to analyze simulated bacterial DNA from the International Space Station (ISS). Before a new crew arrives on station, students must use gel electrophoresis to determine whether a dangerous pathogen lurks before a new crew of astronauts arrives on station. Based on authentic space biotechnology, this lab serves as a fun and engaging introduction to DNA analysis and gel electrophoresis.
Supplied by user:
- Distilled or deionized H₂O (to dissolve TBE buffer)
- Gel electrophoresis system (lab is compatible with Bandit™, blueGel™, or other gel electrophoresis systems)
- Variable volume micropipettes, or fixed-volume minipettes, and tips
Required equipment:
- Micropipettes and tips
- Gel electrophoresis system (e.g., Bandit™, blueGel™, or other gel electrophoresis systems)
Storage notes:
- Reagents must be used within 12 months of shipment
- Agarose and buffer can be stored at room temperature
- Refrigeration is recommended for blue dye samples to prevent evaporation
Accessories information: The learning lab companion kit (catalog no. 470352-026) contains all the consumables (except water) needed for a class of 32 students. These reagents and consumables can also be sourced independently.
Delivery: This lab kit provides enough reagents for eight lab groups: 8 Load Ready™ Strips (tube strips containing pre-aliquoted dye samples); TBE electrophoresis buffer; 8 Agarose Tabs™.