ContactUsLinkComponent
miniPCR® Electrophoresis Forensics Lab: Wrongfully Convicted?
miniPCR® Electrophoresis Forensics Lab: Wrongfully Convicted?
Catalog # 470351-974
Supplier:  AMPLYUS LLC
undefined
miniPCR® Electrophoresis Forensics Lab: Wrongfully Convicted?
Catalog # 470351-974
Supplier:  AMPLYUS LLC
Supplier Number:  KT-1504-01
Specifications
  • Description:
    miniPCR® electrophoresis forensics lab
  • No. of Lab Groups:
    8
  • Cat. no.:
    470351-974
Specifications
About this item
J.M. was convicted of attempted homicide and is currently serving a life sentence.
  • Techniques: Micropipetting, gel electrophoresis
  • Topics: Forensics, genetics, biotechnology
  • Time required: Can be completed in one 45-minute class periods
  • Level: General high school through advanced high school
J.M. has always maintained his innocence, and after years of appeals, the court has approved his request for DNA analysis of the evidence from his case. In this lab students will use gel electrophoresis to examine newly collected DNA evidence from J.M.’s case. Will the DNA evidence prove that J.M. was wrongly convicted, or will it only implicate him further? This lab demonstrates the power that DNA analysis brings to forensic investigations. An optional extension allows students to explore population genetics and the statistical analysis forensic scientists use to evaluate the strength of DNA evidence.
Reagents and consumables supplied by user:
- Agarose (electrophoresis grade)
- DNA stain (e.g., GelGreen™)
- Gel electrophoresis buffer (e.g., 1X TBE)
- Distilled or deionized H₂O (to dilute buffer concentrate – not included in Lab Companion Kit)
- Microtubes
Required equipment:
- Micropipettes and tips
- Gel electrophoresis and fluorescence visualization system
Storage notes:
- Reagents require freezer storage
- Reagents are stable for 12 months when stored in the freezer
Accessories information: The learning lab companion kit (catalog no. 470352-026) contains all the consumables (except water) needed for a class of 32 students. These reagents and consumables can also be sourced independently.
Delivery: The miniPCR Electrophoresis Forensics Lab kit contains reagents for 8 lab groups of up to 4 students each (32 students): Victim DNA sample; J.M. DNA sample; DNA Evidence 1; DNA Evidence 2; DNA molecular weight marker (Fast DNA Ladder 1). Note: Reagents are compatible with blueGel™ or other standard DNA electrophoresis systems.