1321N1 Stable Cells, Human, Multispan
Supplier: MULTISPAN INC
About this item
19 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Documents
MULTISCREEN Stable Cell Lines Expressing GPCRs and Beyond
Application Notes
Multispan MULTISCREEN™ Products and Services
Product Literature
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 76918-522
- 76918-600
- 76918-306
- 76919-178
- 76918-404
- 76919-248
- 76918-738
- 76918-272
- 76918-874
- 76919-100
- 76918-962
- 76918-900
- 76918-638
- 76919-282
- 76918-816
- 76918-946
- 76918-682
- 76918-374
- 76919-018
- Storage conditions:
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
- LN₂ (Below –135 ℃)
Specifications
Product Family Options
Product Information
- Cell FormatSingle Donor/PooledSizeAvailabilityPrice
- 76918-522MULTISCREEN™ Human GPR17 (Short Form) 1321N1 HTS Stable Cell LineMULTISCREEN™ Human GPR17 (Short Form) 1321N1 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1526-3Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76918-522Storage conditionsLN₂ (Below –135 ℃) - 76918-600MULTISCREEN™ Human P2Y11 1321N1 HTS Stable Cell LineMULTISCREEN™ Human P2Y11 1321N1 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1169-3Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76918-600Storage conditionsLN₂ (Below –135 ℃) - 76918-306MULTISCREEN™ Human P2Y12 1321N1 HTS Stable Cell LineMULTISCREEN™ Human P2Y12 1321N1 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1170-3Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76918-306Storage conditionsLN₂ (Below –135 ℃) - 76919-178MULTISCREEN™ Human LPA3 RH7777 HTS Stable Cell LineMULTISCREEN™ Human LPA3 RH7777 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1053-6Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76919-178Storage conditionsLN₂ (Below –135 ℃) - 76918-404MULTISCREEN™ Human LPA3 RH7777 HTS Stable Cell LineMULTISCREEN™ Human LPA3 RH7777 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1053-6AItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76918-404Storage conditionsLN₂ (Below –135 ℃) - 76919-248MULTISCREEN™ Human GPR17 (Long Form) 1321N1 HTS Stable Cell LineMULTISCREEN™ Human GPR17 (Long Form) 1321N1 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1076-3BItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76919-248Storage conditionsLN₂ (Below –135 ℃) - 76918-738MULTISCREEN™ Human LPA5 RH7777 HTS Stable Cell LineMULTISCREEN™ Human LPA5 RH7777 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1145-6Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76918-738Storage conditionsLN₂ (Below –135 ℃) - 76918-272MULTISCREEN™ Human P2Y1 1321N1 HTS Stable Cell LineMULTISCREEN™ Human P2Y1 1321N1 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1160-3Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76918-272Storage conditionsLN₂ (Below –135 ℃) - 76918-874MULTISCREEN™ Human P2Y2 1321N1 HTS Stable Cell LineMULTISCREEN™ Human P2Y2 1321N1 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1161-3Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76918-874Storage conditionsLN₂ (Below –135 ℃) - 76919-100MULTISCREEN™ Human P2Y4 1321N1 HTS Stable Cell LineMULTISCREEN™ Human P2Y4 1321N1 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1162-3Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76919-100Storage conditionsLN₂ (Below –135 ℃) - 76918-962MULTISCREEN™ Human P2Y6 1321N1 HTS Stable Cell LineMULTISCREEN™ Human P2Y6 1321N1 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1164-3Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76918-962Storage conditionsLN₂ (Below –135 ℃) - 76918-900MULTISCREEN™ Human PAR1 HeLa HTS Stable Cell LineMULTISCREEN™ Human PAR1 HeLa HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1207-7Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76918-900Storage conditionsLN₂ (Below –135 ℃) - 76918-638MULTISCREEN™ Human PAR2 HeLa HTS Stable Cell LineMULTISCREEN™ Human PAR2 HeLa HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1208-7Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76918-638Storage conditionsLN₂ (Below –135 ℃) - 76919-282MULTISCREEN™ Human PAR4 HeLa HTS Stable Cell LineMULTISCREEN™ Human PAR4 HeLa HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1210-7Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76919-282Storage conditionsLN₂ (Below –135 ℃) - 76918-816MULTISCREEN™ Human LPA1 RH7777 HTS Stable Cell LineMULTISCREEN™ Human LPA1 RH7777 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1048-6Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76918-816Storage conditionsLN₂ (Below –135 ℃) - 76918-946MULTISCREEN™ Human S1P3 RH7777 HTS Stable Cell LineMULTISCREEN™ Human S1P3 RH7777 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1049-6Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76918-946Storage conditionsLN₂ (Below –135 ℃) - 76918-682MULTISCREEN™ Human LPA2 RH7777 HTS Stable Cell LineMULTISCREEN™ Human LPA2 RH7777 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1050-6Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76918-682Storage conditionsLN₂ (Below –135 ℃) - 76918-374MULTISCREEN™ Human P2Y13 1321N1 HTS Stable Cell LineMULTISCREEN™ Human P2Y13 1321N1 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1139-3Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76918-374Storage conditionsLN₂ (Below –135 ℃) - 76919-018MULTISCREEN™ Human GPR87 RH7777 HTS Stable Cell LineMULTISCREEN™ Human GPR87 RH7777 HTS Stable Cell LineCryopreservedSingle donor2 VialsLog in for availabilityEach$58,965.20Supplier Number: C1140-6Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.76919-018Storage conditionsLN₂ (Below –135 ℃)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



