Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cells
Stable Cells, Mouse
Stable Cells, Mouse
:
MULTISPAN INC
Stable Cells, Mouse
:
MULTISPAN INC
Storage conditions:
LN₂ (Below –135 ℃)
MULTISCREEN Stable Cell Lines Expressing GPCRs and Beyond
Multispan MULTISCREEN™ Products and Services
TDS Stable Cell Line Human Recombinant GPR91 Receptor
TDS Stable Cell Line Human Recombinant mNPS Receptor
TDS Βeta-Arrestin2 Stable Cell Line Mouse Recombinant Mouse CB2 Receptor
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Universal Pipette Tips
: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International