AccuForge® Hobby Blade #11
Catalog # 76717-288
Supplier: Avantor
Specifications
- Coating:Uncoated
- Description:Oiled
- Material:Carbon steel
- Thickness:0.021"
- Packed:1000 blades/case
- Cat. no.:76717-288
- Blade Edge:2 facet
Specifications
About this item
AccuForge #11 Hobby blades are constructed from carbon steel for increased strength, durability and sharpness. Precision acute point for fine cuts.
- Fits all standard #1 Handles
- Excellent for precision cutting and trimming
- Available in bulk pack or 100-packs
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.