AccuForge® Double Edge Blades, 10 Blade Dispenser
Catalog # 76717-296
Supplier: Accutec Blades
Specifications
- Coating:Coated
- Description:Oiled
- Material:Stainless steel
- Thickness:0.004"
- Packed:10 blades/dispenser, 500 dispensers/case
- Cat. no.:76717-296
- Blade Edge:3 facet
Specifications
About this item
AccuForge double edge blades feature a 3-Facet edge for exacting, clean cuts. The MicroCoat® Enrichment technology reduces cut force and helps the blade maintain a sharp effective edge over multiple uses. Safety blade dispenser holds 10 blades.
- Blade edge with MicroCoat® technology enhances durability and sharpness
- 3-facet edge for increased sharpness
- Each dispenser holds 10 blades
AccuForge's MicroCoat® blade enrichment technology adds a layer of protection for each blade helping maintain a sharp effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.