AccuForge® Single Edge Blades, Unbacked, Unwrapped
Supplier: Accutec Blades
Certificates
About this item
AccuForge unbacked single edge blades feature 3-facet edges for sharpness.
- Unbacked blades
- 3-facet edge for increased sharpness
- Available in bulk pack or 100-packs
- Bulk, unwrapped blades for high volume use
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.