AccuForge® Single Edge Blades, Coated, Unwrapped
Catalog # 76717-298
Supplier: Accutec Blades
Specifications
- Coating:Coated
- Description:Oiled
- Material:Stainless steel
- Thickness:0.009"
- Packed:440 blades/tray, 285 trays/crate
- Cat. no.:76717-298
- Blade Edge:3 facet
- Backing Material:Aluminum
Specifications
About this item
AccuForge single edge blades feature 3-facet edges for sharpness. The MicroCoat® Enrichment technology reduces cut force and increases durability.
- Blade edge with MicroCoat® technology enhances durability and sharpness
- Unwrapped blades are ideal for high volume uses
- 3-facet edge for increased sharpness
- Blades are packed in a wooden crate
AccuForge's MicroCoat® blade enrichment technology adds a layer of protection for each blade helping maintain a sharp effective edge over multiple uses.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.