miniPCR® Micropipetting Practice Dyes
470352-002
- Description:miniPCR® micropipetting practice dyes
- Cat. no.:470352-002
Three micropipetting practice dyes: Blue, yellow, and red (5 ml each).
Micropipettes allow scientists to measure small volumes of liquid accurately. Hone your micropipetting skills by practicing with reusable supplies. These Micropipetting Practice Dyes are the same three dyes included with the Micropipetting 101 Learning Lab.
: The Learning Lab Companion Kit (catalog no. 470352-026) contains all the consumables (except water) needed for a class of 32 students. These reagents and consumables can also be sourced independently.
: Includes three micropipetting practice dyes: Blue, yellow, and red (5 ml each).