ContactUsLinkComponent
Merit Dissection Kit with Cassette and 2 Boxes of Blades, Sklar
Catalog # 75833-306
Supplier:  Sklar
undefined
Merit Dissection Kit with Cassette and 2 Boxes of Blades, Sklar
Catalog # 75833-306
Supplier:  Sklar
Supplier Number:  98-1709
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Merit Dissection Kit with Cassette and 2 Boxes of Blades
  • Cat. no.:
    75833-306
  • Supplier No.:
    98-1709
Specifications
About this item
Set for use in laboratory mouse dissections.
Components:
Merit Scalpel Handle - #3
Disposable Stainless Steel Surgical Blades #10
Disposable Scalpel #10
Merit Operating Scissors - S/B, 5-1/2"
Merit Plain Splinter Forceps - 4-1/2"
Merit Webster / Halsey Needle Holder - Smooth, 5-1/4"
Merit Vannas Micro Scissors
Merit Jewelers Forceps
Instrument Cassette 7 x 5-3/4" x 1-1/3"