Amersham ECL Direct Labeling System, Cytiva
CA95038-274L
: Cytiva
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:ECL Direct Labelling Module
- Cat. no.:CA95038-274L
- Application:Labeling 5 µg Nucleic Acid
- Supplier No.:RPN3005
ECL Direct Nucleic Acid Labeling and Detection Systems are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 minute chemical reaction. The resulting probe can be used without purification. Detection is achieved by generation of light via the HRP-catalyzed breakdown of luminol.
- Direct probe labelling in a 10 minute reaction, 1 hour from hybridization to detection with ECL Direct and Hybond™ N+.
- Eliminates handling, waste and regulatory issues associated with use of radioactivity.
- No need to strip blots before reprobing.
- Contains a novel rate enhancer, giving higher sensitivity with very low backgrounds on nylon membranes.
: ECL Direct is particularly useful for applications that require a blot to be reprobed several times and also for screening applications requiring rapid throughput and fast results. Each system includes the following reagents, sufficient for labeling 5 to 10 µg nucleic acid and detecting 2000 to 4000 cm ² of membrane (depending on product ordered): labeling reagent, crosslinker, control DNA, blocking agent, ECL Detection Reagents, and ECL Gold Hybridization Buffer.