- Particle Size:30 - 40 µm
- Description:Superose™ 12 (prep grade)
- Size:125 mL
- pH range:3 - 12
- Matrix:Highly cross-linked agarose
- Fractionation range:1 × 103 - 3 × 105 globular proteins
- Cat. no.:CA95055-854L
- Type:Liquid
- Supplier No.:17-0536-01
Good resolution separations of proteins and other biomolecules according to size, Wide separation range – separations in two molecular weight ranges from 1,000 up to 5 million, Excellent reproducibility and durability
- Excellent reproducibility and durability
Achieve high-resolution separations across exceptionally broad molecular-weight ranges. Low ionic strength eluents (< 0.05 M) can utilize a hydrophobic interaction component to alter the selectivity of Superose for some lipids, peptides, and small aromatic compounds. High performance gel filtration media specially designed for preparative purification of biomolecules. Superose 6 is designed for high-resolution chromatography. Superose 6 prep grade permits easy, efficient scale-up to preparative separations. Superose 12 is for high resolution purification of proteins from 1 × 103 to 3 × 105 molecular weight, while Superose 12 prep grade is useful for preparative purification of these higher molecular weight proteins.