Sepharose™ CL-6B Gel Filtration Base Matrix
Catalog # CA95056-224L
Supplier: Cytiva
CAS Number:
Specifications
- Particle Size:45 - 165 µm
- Description:Sepharose™ CL-6B
- Size:1 L
- pH range:4 - 9
- Matrix:Cross-linked agarose, 6%, spherical
- Fractionation range:1 × 104 - 4 × 106 globular proteins
- Storage temperature:4 - 30 °C
- Description:Sepharose™ CL-6B
- Flow rate:30 cm/h
- Cat. no.:CA95056-224L
- Type:Liquid
- Supplier No.:17-0160-01
Specifications
About this item
Sepharose™ CL-6B is a cross-linked agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix.
- 6% cross-linked agarose gel filtration media
- Proven base matrix for coupling affinity ligands
- Higher degree of crosslinking improve matrix stability
The matrix is not pre-activated and the user performs all steps in coupling.