Order Entry
Canada
ContactUsLinkComponent
 
Mag-Bind® Environmental DNA 96 Kit
 
Mag-Bind® Environmental DNA 96 Kit

 

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Wastewater
    Soil
  • Target molecule:
    DNA

 

Frequently Bought Together