Order Entry
Canada
ContactUsLinkComponent
Mag-Bind® Universal Pathogen Core Kit
Mag-Bind® Universal Pathogen Core Kit
Supplier:  Omega Bio-Tek
Certificates
Mag-Bind® Universal Pathogen Core Kit
Supplier:  Omega Bio-Tek

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Bacteria
  • Target molecule:
    DNA

Specifications

Frequently Bought Together