illustra™ MicroSpin™ G-50 columns, Cytiva
: Cytiva
For rapid purification of labelled DNA (≥20 bases) from unincorporated labelled nucleotides in a volume of 25 to 50 μl using spin column chromatography.
- Columns are prepacked with Sephadex™ G-50 Grade pre-equilibrated in TE buffer with 0,05% Kathon CG/ICP Biocide
- Product recovery will be at least 10% lower than that from ProbeQuant™ G-50 Micro Columns
- Can also be used for buffer exchange/desalting of enzymatic reactions (10 to 100 μl) following heat denaturation and phenol extraction
Note: Not for use with ultrasalt-sensitive automated DNA sequencers.
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Amplified/modified DNA
- Sample type:PCR and other enzymatic reactions
- Target molecule:DNA
- Sample size:100 µL
- Description:illustra™ MicroSpin™ G-50 columns
Frequently Bought Together