- Antibody type:Primary
- Antigen name:Bromodeoxyuridine
- Antigen symbol:BrdU
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:130 µL
- Storage temperature:2...8 °C
- Cat. no.:CA95019-028L
- Supplier No.:RPN202
Anti-Bromodeoxyuridine (Anti-BRdU) antibody is supplied in the same formulation as the monoclonal antibody to BrdU in the Cell Proliferation Kit. The antibody/nuclease mixture is compatible with detection systems that incorporate fluorescently labelled antibodies and biotin-streptavidin detection reagents.
- No need for chemical denaturation
- Detection while maintaining cell morphology
- Double immunocytochemical staining studies can be perfromed
Used in conjunction with RPN20, the reagent allows extended use of the kit where additional amounts of labeling reagent are required, such as in vivo studies.
The antibody contains a nuclease to produce single stranded DNA without the need for chemical denaturation. Cellular morphology is preserved allowing multiple labeling experiments to be carried out. The high-contrast stain produced has sensitivity comparable to autoradiographic methods.
Type: Primary
Antigen: BrdU (Bromodeoxyuridine)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Chemical