Order Entry
Canada
ContactUsLinkComponent
Anti-PEG Mouse Monoclonal Antibody (Biotin) [clone: 5E10E9]
Anti-PEG Mouse Monoclonal Antibody (Biotin) [clone: 5E10E9]
Catalog # 76198-880
Supplier:  Genscript
Anti-PEG Mouse Monoclonal Antibody (Biotin) [clone: 5E10E9]
Catalog # 76198-880
Supplier:  Genscript
Supplier Number:  A01796-100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    PEG
  • Antigen symbol:
    PEG
  • Clonality:
    Monoclonal
  • Clone:
    5E10E9
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgM
  • Western blot:
    Yes
  • Size:
    100 µg
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS, pH 7.4, containing 10 mg/ml BSA and 0.02% sodium azide
  • Storage temperature:
    It is stored lyophilized until use. It remains stable in lyophilized form if stored at −20 °C or below. The reconstituted product stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/ml. For long-term storage, adding glycerol to a final concentration of 50% (v/v) in the reconstituted antibody solution is recommended
  • Shipping temperature:
    Ice
  • Immunogen:
    PEG conjugated to KLH
  • Purification:
    Affinity chromatography
  • Cat. no.:
    76198-880
  • Packaging:
    Vial

Specifications

About this item

GenScript THE™ PEG antibody [Biotin], mAb, mouse detects PEG backbone. The product can detect the variety of PEGs such as PEG40K, PEG20K, PEG5K, PEG12, PEGylated drugs and PEG conjugates.

  • Anti-drug antibodies
  • High specificit
  • High affinity
  • High stability

GenScript's THE™ PEG antibody, mAb, mouse has high affinity for the PEG backbone. The antibody binds to the variety of PEG such as PEG40K, PEG20K, PEG5K, PEG12, PEGylated drugs and PEG conjugates. .

Type: Primary
Antigen: PEG
Clonality: Monoclonal
Clone: 5E10E9
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: