Anti-TIGIT Mouse Monoclonal Antibody [clone: 20A9]
Catalog # 76509-988
Supplier: Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:TIGIT
- Antigen symbol:TIGIT
- Clonality:Monoclonal
- Clone:20A9
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:40 µg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4, containing 0.02% sodium azide
- Storage temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid freeze/thaw cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant TIGIT protein
- Purification:Protein A affinity column
- Cat. no.:76509-988
- Packaging:Vial
Specifications
About this item
The product is specific for human TIGIT.
- Immune checkpoint antibodies
- Positive control for antibody drug early development
- Validate cell line development
- Facilitate assay development
To facilitate this hot immunotherapy research field, GenScript provides immune checkpoint antibodies and immune checkpoint proteins for many kinds of applications. Products for more targets are coming in succession.
.
Type: Primary
Antigen: TIGIT
Clonality: Monoclonal
Clone: 20A9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human