About this item
GenScript’s THE™ PEG Antibody, mAb, Mouse has high affinity for the PEG backbone. The antibody binds to the variety of PEG such as PEG40K, PEG20K, PEG5K, PEG12, PEGylated drugs and PEG conjugates. and nbsp.
- Anti-Drug antibodies
- High specificit
- High affinity
- High stability
GenScript’s THE™ PEG Antibody, mAb, Mouse has high affinity for the PEG backbone. The antibody binds to the variety of PEG such as PEG40K, PEG20K, PEG5K, PEG12, PEGylated drugs and PEG conjugates.
Type: Primary
Antigen: PEG
Clonality: Monoclonal
Clone: 5E10E9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76198-878
- 76198-876
- Antigen symbol:
- PEG
- PEG
- Antigen name:
- PEG
- PEG
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 5E10E9
- 5E10E9
- Host:
- Mouse
- Mouse
- Isotype:
- IgM
- IgM
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- PEG conjugated to KLH
- PEG conjugated to KLH
- Form:
- Lyophilized
- Lyophilized
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml. For long-term storage, adding glycerol to a final concentration of 50% (v/v) in the reconstituted antibody solution is recommended
- Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml. For long-term storage, adding glycerol to a final concentration of 50% (v/v) in the reconstituted antibody solution is recommended
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7.4, containing 0.02% sodium azide
- PBS, pH 7.4, containing 0.02% sodium azide
- Storage temperature:
- The lyophilized product remains stable for up to 1 year at −20 °C from the date of receipt. The reconstituted antibody can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- The lyophilized product remains stable for up to 1 year at −20 °C from the date of receipt. The reconstituted antibody can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Shipping temperature:
- Ice
- Ice