Order Entry
Canada
ContactUsLinkComponent
Anti-Atezolizumab Mouse Monoclonal Antibody [clone: 6B12]
Anti-Atezolizumab Mouse Monoclonal Antibody [clone: 6B12]
Catalog # 76509-926
Supplier:  Genscript
Anti-Atezolizumab Mouse Monoclonal Antibody [clone: 6B12]
Catalog # 76509-926
Supplier:  Genscript
Supplier Number:  A01949-40
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Atezolizumab
  • Antigen symbol:
    Atezolizumab
  • Clonality:
    Monoclonal
  • Clone:
    6B12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Size:
    40 µg
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS, pH 7.4, contains 0.02% sodium azide
  • Storage temperature:
    The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid freeze/thaw cycles
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Atezolizumab
  • Purification:
    Protein A affinity column
  • Cat. no.:
    76509-926
  • Packaging:
    Vial

Specifications

About this item

The product is specific for Atezolizumab. The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript, A01948-40, Anti-Atezolizumab Antibody (10G9), mAb, Mouse.

  • Anti-Idiotype antibodies
  • High specificit
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type: Primary
Antigen: Atezolizumab
Clonality: Monoclonal
Clone: 6B12
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: