Anti-SARS-CoV-2 Nucleocapsid Human Monoclonal Antibody [clone: hIgM2003]
Supplier: Genscript
Certificates
About this item
The product is specific for SARS-CoV-2 Nucleocapsid protein.
- COVID-19 antibodies
- Validated for ELISA, WB, FACS, and SARS-CoV-2 virus neutralization assay
- Control antibodies and various antibody pairs available for ELISA-based detection
- Neutralizing Antibody Standard to calibrate quantitative assay of SARS-CoV-2 neutralizing antibodies in samples
Genscript offers site-specific Neutralizing antibodies, neutralizing antibody standards, and control antibodies.
Type: Primary
Antigen: SARS-CoV-2 Nucleocapsid
Clonality: Monoclonal
Clone: hIgM2003
Conjugation:
Epitope:
Host: Human
Isotype: IgM
Reactivity:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76510-162
- 76510-164
- Antigen symbol:
- SARS-CoV-2 Nucleocapsid
- SARS-CoV-2 Nucleocapsid
- Antigen name:
- SARS-CoV-2 Nucleocapsid
- SARS-CoV-2 Nucleocapsid
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- hIgM2003
- hIgM2003
- Host:
- Human
- Human
- Isotype:
- IgM
- IgM
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant SARS-CoV-2 Nucleocapsid Protein
- Recombinant SARS-CoV-2 Nucleocapsid Protein
- Form:
- Liquid
- Liquid
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7.4, containing 0.02% sodium azide
- PBS, pH 7.4, containing 0.02% sodium azide
- Storage temperature:
- Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed Avoid repeated freezing and thawing cycles
- Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed Avoid repeated freezing and thawing cycles
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together