Anti-MBP Tag Mouse Monoclonal Antibody [clone: 5B10B11]
76509-806
- Antibody type:Tag
- Antigen name:MBP Tag
- Antigen symbol:MBP Tag
- Clonality:Monoclonal
- Clone:5B10B11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Size:40 µg
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4, containing 0.02% sodium azide
- Storage temperature:The antibody is stable in lyophilized form if stored at −20 °C or below. The reconstituted antibody can be stored for 2 - 3 weeks at 2 - 8 °C. For long term storage, aliquot and store at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:Purified recombinant MBP fusion protein
- Purification:Protein A affinity column
- Cat. no.:76509-806
- Packaging:Vial
GenScript MBP-tag Antibody, mAb, Mouse specifically reacts with fusion proteins containing MBP epitope tags.
- Comprehensive pool
- against 24 tags
- Available in gram level bulk quantities
- Various conjugates for hot tags detection
GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis,
immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.
Type: Tag
Antigen: MBP Tag
Clonality: Monoclonal
Clone: 5B10B11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: