Order Entry
Canada
ContactUsLinkComponent
 
Anti-PSA Mouse Monoclonal Antibody [clone: 8A9B8]
 :  Genscript
 
Anti-PSA Mouse Monoclonal Antibody [clone: 8A9B8]
 :  Genscript
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Catalog No:
  • 76509-816
  • 76509-818
  • Antigen symbol:
  • PSA
  • PSA
  • Antigen name:
  • PSA
  • PSA
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 8A9B8
  • 8A9B8
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Immunogen:
  • human total PSA purified from semenal plasma
  • human total PSA purified from semenal plasma
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Protein A affinity column
  • Protein A affinity column
  • Concentration:
  • 0.5 mg/ml
  • 0.5 mg/ml
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS buffer, pH 7.4, containing 0.02% sodium azide
  • PBS buffer, pH 7.4, containing 0.02% sodium azide
  • Storage temperature:
  • For long term storage, aliquot and store at −20 °C or below. Avoid repeated freezing and thawing cycles
  • For long term storage, aliquot and store at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Shipping temperature:
  • Ice
  • Ice

 

Frequently Bought Together