CyBio® Selma Semi-Automatic Pipetes 96/384
Supplier: Analytik Jena US
Certificates
About this item
Precise and semi-automatic pipettor with 96 or 384 pipetting channels for easy and fast filling of 96- and 384-well microplates without the use of a computer. Applications like replication and reformatting, medium exchange as well as serial dilution can be executed reproducible and convenient.
- Fast and precise processing of 96- and 384-well microplates without the need for programming
- Compact footprint and fits on any lab bench as well as into most laminar flow hoods, Simple and intuitive operation via user guided methods
- Easy and fast tip changing with ready to use CyBio Tip Trays
- Error-free and reproducible results due to 96 or 384 parallel pistons and proven tip sealing principle
- Memory function and automatic use of pipetting heights and liquid handling parameters
- Applications like the replication or the reformatting of whole microplates can be executed in seconds
- Save your valuable lab space, Simplify your pipetting tasks, Comfortable and Precise in every detail
No more strenuous and monotonous pipetting by hand, no more empty or inaccurate filling of wells and no more repetition of tests because of pipetting inaccuracies - CyBio® SELMA enables a safe and error free transfer of 96 or 384 samples in a single step.
Customers Who Bought This Also Bought