- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:BTC
- Description:Mouse BTC ELISA kit
- Size:96 tests
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Mouse BTC ELISA kit exhibits high specificity and excellent specificity for the detection of mouse BTC. No significant cross-reactivity or interference between BTC and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:7.813 - 500 pg/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:4.688 pg/ml
- Regulatory status:RUO
- Cat. no.:76735-562
Mouse BTC ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse BTC in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 7.813 - 500 pg/ml
- Sensitivity: 4.688 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Mouse BTC ELISA kit (A73784) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse BTC in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for BTC has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the BTC present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-BTC Antibody, which binds the captured BTC present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of BTC captured in each well. The concentration of BTC can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.