Mouse Inhibin B ELISA Kit
Mouse Inhibin B ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse Inhibin B in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 15.625 - 1000 pg/ml
- Sensitivity: 9.375 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Mouse Inhibin B ELISA kit (A76814) employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse Inhibin B in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Inhibin B antigen. During the incubation, Inhibin B present in the samples or standards competes with the fixed amount of immobilized Inhibin B for binding sites on the Biotinylated Anti-Inhibin B Antibody. The more Inhibin B present in a sample or standard, the less Biotinylated Anti-Inhibin B Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Inhibin B Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Inhibin B present in each sample or standard. The concentration of Inhibin B can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Documents
- Description:Mouse Inhibin B ELISA kit
- Size:96 tests
- Primary antibody reactivity:Mouse
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Sample volume:50 μl
- Detection range:15.625 - 1000 pg/ml
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Competitive
- Time to Results:4 h 30 min
- Cross reactivity:Mouse Inhibin B ELISA kit exhibits high specificity and excellent specificity for the detection of mouse Inhibin B. No significant cross-reactivity or interference between Inhibin B and analogues was observed.
- Assay Principle:Quantitative
- Cat. No.:76739-846
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:9.375 pg/ml
- Format:Pre-coated
- Target protein:Inhibin B
- Regulatory status:RUO
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



