Peroxidase ELISA Kit
76738-956
:
- Assay duration:Multiple steps
- Assay Type:Indirect
- Format:Pre-coated
- Primary antibody reactivity:Other
- Target protein:Peroxidase
- Description:Peroxidase ELISA kit
- Size:96 tests
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:3.12 - 200 pmol/ml
- Storage temperature:4 °C
- Sensitivity:1.875 pg/ml
- Regulatory status:RUO
- Cat. no.:76738-956
Peroxidase ELISA kit is a enzyme catalysis Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of Peroxidase in serum, plasma, tissue homogenates, and other biological fluids.