Human Hepatitis C Virus IgM ELISA Kit
Catalog # 76733-266
Supplier: ANTIBODIES.COM LLC
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Indirect
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Hepatitis C Virus IgM
- Description:Human Hepatitis C virus IgM ELISA kit
- Size:96 tests
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Qualitative
- Shelf life:Store for 6 months at 4 °C
- Storage temperature:4 °C
- Regulatory status:RUO
- Cat. no.:76733-266
Specifications
About this item
Human Hepatitis C Virus IgM ELISA kit is a trapping Enzyme-Linked Immunosorbent Assay designed for the in vitro qualitative determination of human Hepatitis C Virus IgM in serum, plasma, tissue homogenates, and other biological fluids.