Order Entry
Canada
Orders LinkContactUsLinkComponent
Rat eNOS ELISA Kit

Rat eNOS ELISA Kit

Rat eNOS ELISA kit, 96 tests
Catalog # 76730-236
Supplier:   Antibodies.com
Supplier #:  A74497-96
Rat eNOS ELISA Kit
Catalog # 76730-236
Supplier:  Antibodies.com
CAS Number:  
Rat eNOS ELISA Kit
Rat eNOS ELISA Kit

Rat eNOS ELISA kit, 96 tests

CAS Number:  
Price
$1,071.04
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Rat eNOS ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat eNOS in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 15.625 to 1000 pg/ml
  • Sensitivity: 9.375 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat eNOS ELISA Kit (A74497) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat eNOS in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for eNOS has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the eNOS present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-eNOS Antibody, which binds the captured eNOS present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of eNOS captured in each well. The concentration of eNOS can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Description:
    Rat eNOS ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Rat
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    100 μl
  • Detection range:
    15.625 - 1000 pg/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Rat eNOS ELISA Kit exhibits high specificity and excellent specificity for the detection of rat eNOS. No significant cross-reactivity or interference between eNOS and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76730-236
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    9.375 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    eNOS
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.