Human Ovalbumin Specific IgG ELISA Kit
76741-992
:
- Assay duration:Multiple steps
- Assay Type:Indirect
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Ovalbumin Specific IgG
- Description:Human ovalbumin specific IgG ELISA kit
- Size:96 tests
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:0.313 - 20 ng/ml
- Storage temperature:4 °C
- Sensitivity:0.188 ng/ml
- Regulatory status:RUO
- Cat. no.:76741-992
Human Ovalbumin Specific IgG ELISA kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of human Ovalbumin Specific IgG in serum, plasma, tissue homogenates, and other biological fluids.