Order Entry
Canada
ContactUsLinkComponent
Bovine Visfatin ELISA Kit
Bovine Visfatin ELISA Kit
Catalog # 76735-972
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Bovine Visfatin ELISA Kit
Catalog # 76735-972
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A87015-96
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Bovine
  • Target protein:
    Visfatin
  • Description:
    Bovine Visfatin ELISA kit
  • Size:
    96 tests
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Bovine Visfatin ELISA Kit exhibits high specificity and excellent specificity for the detection of bovine Visfatin. No significant cross-reactivity or interference between Visfatin and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    1.563 - 100 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    0.938 ng/ml
  • Regulatory status:
    RUO
  • Cat. no.:
    76735-972

Specifications

About this item

Bovine Visfatin ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of bovine Visfatin in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 1.563 to 100 ng/ml
  • Sensitivity: 0.938 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Bovine Visfatin ELISA Kit (A87015) employs the sandwich enzyme immunoassay technique for the quantitative measurement of bovine Visfatin in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Visfatin has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Visfatin present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Visfatin Antibody, which binds the captured Visfatin present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Visfatin captured in each well. The concentration of Visfatin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.