Order Entry
Canada
ContactUsLinkComponent
Rat IL-15RA ELISA Kit
Rat IL-15RA ELISA Kit
Catalogue # 76732-732
Fournisseur:  ANTIBODIES.COM LLC
Numéro CAS:  
undefined
Rat IL-15RA ELISA Kit
Catalogue # 76732-732
Fournisseur:  ANTIBODIES.COM LLC
Supplier Number:  A303706-96
Numéro CAS:  

Spécifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    IL-15RA
  • Description:
    Rat IL-15RA ELISA kit
  • Size:
    96 tests
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross reactivity:
    Rat IL-15RA ELISA Kit exhibits high specificity and excellent specificity for the detection of rat IL-15RA. No significant cross-reactivity or interference between IL-15RA and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    31.25 - 2000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    18.75 pg/ml
  • Regulatory status:
    RUO
  • Cat. no.:
    76732-732

Spécifications

À propos de cet article

Rat IL-15RA ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat IL-15RA in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Rat IL-15RA ELISA kit (A303706) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat IL-15RA in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for IL-15RA has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IL-15RA present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-IL-15RA Antibody, which binds the captured IL-15RA present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of IL-15RA captured in each well. The concentration of IL-15RA can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.