Human ROR2 ELISA Kit
Human ROR2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ROR2 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human ROR2 ELISA kit (A247186) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human ROR2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for ROR2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the ROR2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-ROR2 Antibody, which binds the captured ROR2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of ROR2 captured in each well. The concentration of ROR2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
- Description:Human ROR2 ELISA kit
- Size:96 tests
- Primary antibody reactivity:Human
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Sample volume:100 μl
- Detection range:0.156 - 10 ng/ml
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Sandwich
- Time to Results:4 h 30 min
- Cross reactivity:Human ROR2 ELISA Kit exhibits high specificity and excellent specificity for the detection of human ROR2. No significant cross-reactivity or interference between ROR2 and analogues was observed.
- Cat. No.:76731-394
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:0.094 ng/ml
- Format:Pre-coated
- Target protein:ROR2
- Regulatory status:RUO



