- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:SLC26A4
- Description:Rat SLC26A4 ELISA kit
- Size:96 tests
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross reactivity:Rat SLC26A4 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat SLC26A4. No significant cross-reactivity or interference between SLC26A4 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:0.313 - 20 ng/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:0.188 ng/ml
- Regulatory status:RUO
- Cat. no.:76744-218
Rat SLC26A4 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat SLC26A4 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Rat SLC26A4 ELISA kit (A247368) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat SLC26A4 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for SLC26A4 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SLC26A4 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-SLC26A4 Antibody, which binds the captured SLC26A4 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of SLC26A4 captured in each well. The concentration of SLC26A4 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.