Order Entry
Canada
Orders LinkContactUsLinkComponent
Dehydroepiandrosterone ELISA Kit | Avantor

Dehydroepiandrosterone ELISA Kit

Dehydroepiandrosterone ELISA kit, 96 tests
Catalog # 76737-088
Supplier:   Antibodies.com
Fournisseur #:  A74191-96
Dehydroepiandrosterone ELISA Kit
Catalog # 76737-088
Supplier:  Antibodies.com
CAS Number:  
Dehydroepiandrosterone ELISA Kit
Dehydroepiandrosterone ELISA Kit

Dehydroepiandrosterone ELISA kit, 96 tests

CAS Number:  
Price
$1,071.04
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Dehydroepiandrosterone ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Dehydroepiandrosterone in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 0.156 - 10 ng/ml
  • Sensitivity: 0.094 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Dehydroepiandrosterone ELISA kit (A74191) employs the competitive enzyme immunoassay technique for the quantitative measurement of universal Dehydroepiandrosterone in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Dehydroepiandrosterone antigen. During the incubation, Dehydroepiandrosterone present in the samples or standards competes with the fixed amount of immobilized Dehydroepiandrosterone for binding sites on the Biotinylated Anti-Dehydroepiandrosterone Antibody. The more Dehydroepiandrosterone present in a sample or standard, the less Biotinylated Anti-Dehydroepiandrosterone Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Dehydroepiandrosterone Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Dehydroepiandrosterone present in each sample or standard. The concentration of Dehydroepiandrosterone can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Spécifications

Spécifications

  • Description:
    Dehydroepiandrosterone ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Other
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    50 μl
  • Detection range:
    0.156 - 10 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Competitive
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Dehydroepiandrosterone ELISA kit exhibits high specificity and excellent specificity for the detection of universal Dehydroepiandrosterone. No significant cross-reactivity or interference between Dehydroepiandrosterone and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76737-088
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0.094 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    Dehydroepiandrosterone
  • Regulatory status:
    RUO

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.